missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TARS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577090
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Threonyl-tRNA synthetase belongs to the class-II aminoacyl-tRNA synthetase family.
Specifications
| TARS | |
| Polyclonal | |
| Unconjugated | |
| TARS1 | |
| D15Wsu59e; hypothetical protein LOC510075; MGC9344; TARS; TARS1; threonine tRNA ligase 1, cytoplasmic; threonine--tRNA ligase; threonine--tRNA ligase 1, cytoplasmic; threonine--tRNA ligase 1, cytoplasmic; threonine--tRNA ligase, cytoplasmic; threonine--tRNA ligase, cytoplasmic; Threonyl-tRNA synthetase; threonyl-tRNA synthetase 1; threonyl-tRNA synthetase, cytoplasmic; ThrRS; wu:fb07c05; wu:fb39c12; zgc:92586; zgc:92586 protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 110960, 294810, 6897 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P26639, Q5XHY5, Q9D0R2 | |
| TARS1 | |
| Recombinant full length Human TARS. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction