missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TARS Polyclonal Antibody

Product Code. 13493157
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13493157 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13493157 Supplier Invitrogen™ Supplier No. PA529040

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa. Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (91%), Xenopus laevis (92%), Chicken (93%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Threonyl-tRNA synthetase belongs to the class-II aminoacyl-tRNA synthetase family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TARS
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene TARS1
Gene Accession No. P26639
Gene Alias D15Wsu59e; hypothetical protein LOC510075; MGC9344; TARS; TARS1; threonine tRNA ligase 1, cytoplasmic; threonine--tRNA ligase; threonine--tRNA ligase 1, cytoplasmic; threonine--tRNA ligase 1, cytoplasmic; threonine--tRNA ligase, cytoplasmic; threonine--tRNA ligase, cytoplasmic; Threonyl-tRNA synthetase; threonyl-tRNA synthetase 1; threonyl-tRNA synthetase, cytoplasmic; ThrRS; wu:fb07c05; wu:fb39c12; zgc:92586; zgc:92586 protein
Gene Symbols TARS1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ThrRS. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6897
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.