Learn More
Thermo Scientific™ Target Polyspring Insert
Polyspring inserts for 9mm HPLC and GC autosampler vials are available in a variety of configurations with usable volumes from 160μL to 3.5mL
Brand:  Thermo Scientific™ C4010-630TM
Product Code. 12433668
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Pulled point inserts are an economical choice for noncritical applications
  • Nominal O.D. of 6mm to fit Target wide-opening vials
TRUSTED_SUSTAINABILITY
Specifications
Specifications
300 μL
100