missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TARC Recombinant Rabbit Monoclonal Antibody (2F6L8)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557890
This item is not returnable.
View return policy
Description
Immunogen sequence: ARGTNVGREC CLEYFKGAIP LRKLKTWYQT SEDCSRDAIV FVTVQGRAIC SDPNNKRVKN AVKYLQSLER S.
TARC ligand (CCL17) belongs to the CC chemokine family. It is a small cytokine and the gene is located on q arm of chromosome 16, in humans, along with other chemokines. TARC specifically binds to T-cells and interacts with chemokine receptors CCR4 and CCR8. It plays an important role in T cell development in thymus as well as in trafficking and activation of mature T cells.
Specifications
| TARC | |
| Recombinant Monoclonal | |
| 1.86 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q92583 | |
| CCL17 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 24-94 of human CCL17 (NP_002978.1). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 2F6L8 | |
| Unconjugated | |
| CCL17 | |
| A-152E5.3; ABCD-2; CC chemokine TARC; C-C motif chemokine; C-C motif chemokine 17; C-C motif chemokine ligand 17; CCL17; chemokine (C-C motif) ligand 17; SCYA17; Scya17l; small inducible cytokine subfamily A (Cys-Cys), member 17; small inducible cytokine subfamily A17; small-inducible cytokine A17; T cell-directed CC chemokine; TARC; Thymus and activation-regulated chemokine | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6361 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction