missing translation for 'onlineSavingsMsg'
Learn More

TARBP1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18396805
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396805 100 μg 100µL
18339223 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18396805 Supplier Novus Biologicals Supplier No. NBP316035100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TARBP1 Polyclonal antibody specifically detects TARBP1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TARBP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias EC 2.1.1.-, FLJ30482, TAR (HIV-1) RNA binding protein 1, TAR RNA loop binding protein, TAR RNA-binding protein 1, TAR RNA-binding protein of 185 kDa, tardbp1, TRM3probable methyltransferase TARBP1, TRP-185TAR (HIV) RNA binding protein 1, TRP185TAR (HIV) RNA-binding protein 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1322-1621 of human TARBP1 (NP_005637.3).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 6894
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.