missing translation for 'onlineSavingsMsg'
Learn More

TARBP1 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18410527 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410527 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410527 Supplier Novus Biologicals Supplier No. NBP176400G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TARBP1 Polyclonal antibody specifically detects TARBP1 in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TARBP1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias EC 2.1.1.-, FLJ30482, TAR (HIV-1) RNA binding protein 1, TAR RNA loop binding protein, TAR RNA-binding protein 1, TAR RNA-binding protein of 185 kDa, tardbp1, TRM3probable methyltransferase TARBP1, TRP-185TAR (HIV) RNA binding protein 1, TRP185TAR (HIV) RNA-binding protein 1
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide from near the center of human TRBP1. The immunogen is located within amino acids 1310 - 1360 of TRBP1. Amino Acid Squence: HGAGNAKKNWQRIQEHF
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 6894
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.