missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ siRNA Assay
Shop All Applied Biosystems Products
Click to view available options
No. of Reactions:
25 Reactions RT primer/75 Reactions TaqMan assay
2900 Reactions
50 Reactions RT primer/150 Reactions TaqMan assay
750 Reactions
Unit Size:
Each
Description
TaqMan™ siRNA Assays, Made-to-Order
Specifications
Specifications
| Concentration | 60X (Assay), 60X (RT Primer) |
| Description | TaqMan™ siRNA Assay, 1 x 725μL RT primer/1 x 967μL TaqMan™ Assay 2900 rxns, Large, 60X RT Primer/60X TaqMan™ Assay Concentration, Store at –15 to –25°C |
| Product Type | TaqMan™ siRNA Assay |
| Quantity | 1 Each |
| No. of Reactions | 2900 Reactions |
| Volume | 1 x 725μL RT primer/1 x 967μL TaqMan™ assay |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction