missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ siRNA Assay

Product Code. 15561074 Shop All Applied Biosystems Products
Change view
Click to view available options
No. of Reactions:
25 Reactions RT primer/75 Reactions TaqMan assay
2900 Reactions
50 Reactions RT primer/150 Reactions TaqMan assay
750 Reactions
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
15561074 25 Reactions RT primer/75 Reactions TaqMan assay Each
15571074 50 Reactions RT primer/150 Reactions TaqMan assay Each
15581074 750 Reactions Each
15591074 2900 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15561074 Supplier Applied Biosystems™ Supplier No. 4440877
  • ONLINE EXCLUSIVE PRICE
    £274.00
    £210.98 / Pack of 1
    Save £63.02
    23% Off
View all promotions
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

TaqMan™ siRNA Assay

TaqMan™ siRNA Assays, Made-to-Order
TRUSTED_SUSTAINABILITY

Specifications

Concentration 20X (Assay), 5X (RT Primer)
Description TaqMan™ siRNA Assay, 1 x 75μL RT primer/1 x 75μL TaqMan™ Assay, 25 rxns RT Primer/75 rxns TaqMan™ Assay, Extra Small, 5X RT primer/20X TaqMan™ Assay Concentration, Store at –15 to –25°C
Product Type TaqMan™ siRNA Assay
Quantity 1 Each
No. of Reactions 25 Reactions RT primer/75 Reactions TaqMan assay
Volume 1 x 75μL RT primer/1 x 75μL TaqMan™ assay

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.