missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ SARS-CoV-2, Flu A/B, RSV RNA Control

Product Code. 17296503
Click to view available options
Quantity:
10 x 10 μL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17296503

Brand: Applied Biosystems™ 956126

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The TaqMan SARS-CoV-2, Flu A/B, RSV RNA Control contains targets specific to the SARS-CoV-2, influenza A/B, and RSV genomic regions targeted by the TaqMan SARS-CoV-2, FluA/B RSV RT-PCR Assay Kit.

The TaqMan™ SARS-CoV-2, Flu A/B, RSV RNA Control is recommended for use with the TaqMan SARS-CoV-2, FluA/B RSV RT-PCR Assay Kit as a positive control to ensure reaction setup and reagent integrity when interpreting test results.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at ≤-70°C.
Format Tube(s)
For Use With TaqMan™ SARS-CoV-2, Flu A/B, RSV RT-PCR Assay Kit
Product Line TaqMan
Quantity 10 x 10 μL
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.