missing translation for 'onlineSavingsMsg'
Learn More

TaqMan™ Roundup Ready Soya Quantification Kit

Product Code. 13502550 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
2 x 48 rxn(s)
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13502550 2 x 48 rxn(s) Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13502550 Supplier ABI Applied Biosystems Supplier No. 4466335

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TaqMan™ Roundup Ready Soya Quantification Kit

The TaqMan™ Roundup Ready Soya Quantification Kit is a food safety testing kit used to determine the percentage of Roundup Ready soya against the total amount of soya present in a DNA sample. This real-time PCR kit contains all reagents (except for the template DNA) required for the quantification of Roundup Ready soya. The benefits of the TaqMan™ Roundup Ready Soya Quantification Kit over other commercially available products include:

  • Efficiency—detection of small DNA fragments through use of Taqman™ MGB probes
  • Sensitivity—quantification limit of 20 DNA copies and detection limit of 3 DNA copies
  • Flexibility—adapted to processed DNAs

    The TaqMan™ Roundup Ready Soya Quantification Kit consists of plasmid-containing targets for the endogenous soy gene, known as lectin, and for the transgenic event (GTS-40-3-2) present in Roundup Ready soy. This soy accounts for approximately 80% of all soy cultivated worldwide and is considered to be the most widespread transgenic event.

    The TaqMan™ Roundup Ready Soya Quantification Kit is optimized for use in all Applied Biosystems™ real-time PCR instruments.

    The development and manufacture of this kit occurs through a partnership with IMEGEN (Instituto de Medicina Genomica), which has a long history of GMO research and development.

    Other related products include:
    GMO Extraction Kit
    TaqMan™ GMO Screening Kit
    TaqMan™ GMO Maize Quantification Kit
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Roundup Ready™ soya master mix (green disc), soya master mix (purple disc), general master mix (white disc), Roundup Ready™ soya standard (green cap). –20/4°C
    Description TaqMan™ Roundup Ready Soya Quantification Kit, 2 x 48 rxns, Food Sample Type, Lectin, GTS-40-3-2 Target Gene, Primer-Probe Detection Method, For Food Safety Testing
    Detection Method Primer-probe
    Product Type TaqMan™ Roundup Ready Soya Quantification Kit
    Quantity 2 x 48 rxn(s)
    Target Lectin, GTS-40-3-2
    For Use With (Application) Food and Beverage
    No. of Reactions 96 Reactions
    Sample Type Food
    Size (Final Product) 2 x 48 rxn(s)

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.