missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Protein Assays Open Kit

Product Code. 12057799 Shop All Applied Biosystems Products
Click to view available options
Quantity:
4,000 reactions
Unit Size:
4000 reactions
This item is not returnable. View return policy

Product Code. 12057799

Brand: Applied Biosystems™ 4453745

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Sensitive Detection — directly measure in vitro protein-protein interactions with only a few thousand cells.

  • Small Sample Size - use up to 500 times less sample input compared to traditional methods
  • Sensitive Detection - directly measure in vitro protein-protein interactions with only a few thousand cells
  • Revolutionary - directly correlate mRNA and/or microRNA to protein
  • Simple Workflow - 1-step lysis sample preparation, universal protocol
  • Quantitative - protein marker expression fold changes detected
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The TaqMan™ Protein Assays Open Kit contains the TaqMan™ Protein Assays Oligo Probe Kit (4448549) and the TaqMan™ Protein Assays Buffer Kit (4448571)for making TaqMan™ Protein Assay probes.
Detection Method Primer-probe
GC-Rich PCR Performance Low
PCR Method qPCR
Reaction Speed Fast
Technique Proximity Ligation Assay
For Use With (Equipment) 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, StepOnePlus™, Fast Mode
Green Features Sustainable packaging
Label or Dye FAM
Passive Reference Dye ROX (Pre-mixed)
Product Line TaqMan
Product Type Protein Assays Open Kit
Quantity 4,000 reactions
Target Specificity Not Target-Specific
Hot Start No
No. of Reactions 4000 Reactions
Sample Type Tissue, Cells
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.