missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Protein Assays Fast Master Mix

Product Code. 11330650 Shop All Applied Biosystems Products
Click to view available options
Quantity:
500 Reactions
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11330650

Brand: Applied Biosystems™ 4448616

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

TaqMan™ Protein Assays Fast Master Mix

The enzyme concentration, ionic strength, and the pH of the TaqMan™ Protein Assays Fast Master Mix are optimized for the amplification of ligated oligonucleotide target sequences prepared using TaqMan™ Protein Assays reagents. The DNA polymerase included in the master mix is designed for maximum PCR efficiency and specificity when used with the ligation chemistry.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage TaqMan™ Protein Assays Fast Master Mix is optimized for the amplification of ligated oligonucleotide target sequences prepared using TaqMan™ ProteinAssays reagents
Detection Method Primer-probe Detection
Product Line TaqMan
Sample Type Cells, Tissue
Specificity Not Target-Specific
For Use With (Application) Proximity Ligation Assay
For Use With (Equipment) 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, StepOnePlus™, Fast Mode
Label or Dye FAM™
Quantity 500 Reactions
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.