missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Protein Assays Fast Master Mix (2X)

Product Code. 11501296 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
100 reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11501296 100 reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11501296 Supplier Applied Biosystems™ Supplier No. 4400088

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TaqMan™ Protein Assays Fast Master Mix (2X)

The enzyme concentration, ionic strength, and the pH of the TaqMan™ Protein Fast Master Mix are optimized for the amplification of ligated oligonucleotide target sequences prepared using TaqMan™ Protein Assay reagents. The DNA polymerase included in the master mix is designed for maximum PCR efficiency and specificity when used with the ligation chemistry.

Running a complete TaqMan™ Protein Assay (pre-designed or made with the Open Kit) requires a Core Reagents Base Kit and Master Mix Kit.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Store kit at 2-8°C.
Detection Method Primer-probe
PCR Method qPCR
Polymerase AmpliTaq Fast DNA Polymerase
Reaction Speed Fast
Technique Proximity Ligation Assay
For Use With (Equipment) 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, AB 7900 HT 384-Well System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode
Passive Reference Dye ROX (Pre-mixed)
Product Line TaqMan
Quantity 100 reactions
Shipping Condition Wet Ice
Target Specificity Not Target-Specific
For Use With (Application) Cell Analysis
Hot Start No
No. of Reactions 100 reactions
Sample Type Cells
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.