missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ OpenArray™ Real-Time PCR Master Mix

Product Code. 15591114 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1.5 mL
Unit Size:
1.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15591114 1.5 mL 1.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15591114 Supplier Applied Biosystems™ Supplier No. 4462159
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ OpenArray™ Real-Time PCR Master Mix is optimized for use with TaqMan™ OpenArray™ gene expression panels, and yields reproducible, specific, and sensitive results.

  • Hot start chemistry to provide room temperature stability
  • Optimized for use with TaqMan™ OpenArray™ gene expression panels yielding reproducible, specific, and sensitive results
  • For use with OpenArray™ MicroRNA panels, OpenArray™ gene expression pathway panels, and other gene expression applications on the OpenArray™ format
  • Ready for use with no additional additives required

Order Info

Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -5 to -30°C until first use, then at 2–8°C.
Detection Method Primer-probe
Form Liquid
Format Tube
PCR Method 2-step RT-qPCR
Reaction Speed Standard
Technique Fluorescence Intensity
For Use With (Equipment) OpenArray™
Product Line OpenArray, TaqMan
Product Type qPCR Master Mix
Quantity 1.5 mL
Shipping Condition Dry Ice
Target Specificity Target-Specific
For Use With (Application) miRNA analysis
Hot Start Built-In Hot Start
No. of Reactions 10 Panels
Sample Type Total RNA
Volume 1.5 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.