missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ OpenArray™ Genotyping Master Mix

Product Code. 15569992 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15569992 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15569992 Supplier Applied Biosystems™ Supplier No. 4404846
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This 2X concentrated mix is optimized for SNP genotyping using TaqMan™ OpenArray™ SNP Genotyping Plates.

  • PCR Method: qPCR
  • Built-In Hot Start Primer-Probe Compatability: TaqMan™

Order Info

Shipping Conditions: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Includes TaqMan™ OpenArray™ Genotyping Master Mix, with AmpliTaq Gold™ DNA Polymerase UP (Ultra Pure), dNTPs, and optimized mix components. This pack contains one 5 mL bottle, sufficient for 32 plates. Store at 2–8°C.
Detection Method Primer-probe
PCR Method qPCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
Technique SNP Genotyping (Real-Time PCR-Based)
For Use With (Equipment) OpenArray™
Genotyping Target Drug Metabolizing Enzymes, SNPs (Known)
Passive Reference Dye ROX (Pre-mixed)
Product Line OpenArray, TaqMan
Product Type Genotyping Master Mix
Quantity 1 kit
Shipping Condition Wet Ice
For Use With (Application) Genotyping
Fidelity (vs. Taq) 2 X
Hot Start Built-In Hot Start
Sample Type DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.