missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Non-Coding RNA Assay
Shop All Applied Biosystems Products
Click to view available options
Description:
Extra small scale
Small scale
Unit Size:
Each
Description
- TaqMan™ Non-coding RNA Assays provide a highly reliable way to measure expression of long non-coding transcripts.
- These assays are based on TaqMan™ Assays technology to help provide the high specificity and sensitivity you need for quantification and validation studies.
Specifications
Specifications
| Content And Storage | Store in freezer (-5° to -30°C). |
| Description | Small scale |
| Form | Liquid |
| Product Line | TaqMan |
| Quantity | small scale |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction