missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Non-coding RNA Assay
Shop All Applied Biosystems Products

Click to view available options
Quantity:
L (2900 rxns), made to order
M (750 rxns), made to order
S (360 rxns), made to order
Unit Size:
Each
Specifications
Specifications
| Content And Storage | 1 tube containing a 20X (S and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers). Store at -15 to -25°C. |
| For Use With (Application) | Real Time PCR (qPCR) |
| Green Features | Sustainable packaging |
| No. of Reactions | 750 reactions |
| Product Line | TaqMan |
| Quantity | M (750 rxns), made to order |
| Shipping Condition | Room Temperature |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction