missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Mutation Detection Assay with castPCR™ Technology
GREENER_CHOICE

Product Code. 13345384 Shop All Applied Biosystems Products
Click to view available options
Quantity:
Gene Reference (75 reactions), Inventoried
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 13345384

Brand: Applied Biosystems™ 4465807

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Powered by competitive allele-specific TaqMan™ PCR (castPCR™ Technology) to detect and measure somatic mutations in genes associated with cancer research.

The castPCR technology is highly specific and sensitive, and can detect rare amounts of mutated DNA in a sample that contains large amounts of normal, wild-type DNA. These assays are compatible with different sample types, such as cell lines, formalin-fixed paraffin-embedded (FFPE) tissue samples, and fresh frozen tissue samples.

Competitive, allele-specific TaqMan PCR utilizes an allele-specific primer for mutant allele detection that competes with an MGB blocker oligonucleotide to suppress the wild-type background.

  • Higher specificity: designed to block the wild-type and amplify only the mutant
  • Higher sensitivity: designed to detect somatic mutations down to 1 cancer cell in 1000 normal cells
  • Faster workflow: 3 hours from sample to result
  • Detect multiple types of mutations: detect single- and multiple-nucleotide mutations and insertion/deletions (indels)
  • Wide dynamic range and excellent PCR efficiency: assays demonstrate at least 4 logs of dynamic range and an average efficiency of 100% ± 10%
  • Fast, simple workflow: like other TaqMan Assays, typically require 3 hours from sample to results, with minimum hands-on time

Order Info

Expiration date 2 years from the date of manufacturer. Shipping Conditions: Ambient Temperature

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10X
Content And Storage 1 tube containing a 10X mix of unlabeled PCR primers and a TaqMan MGB probe

Store at -15 to -25°C.

Detection Method Primer-probe
Form Frozen
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Reaction Speed Fast
Product Line TaqMan
Product Type Mutation Detection Assay
Quantity Gene Reference (75 reactions), Inventoried
Shipping Condition Room Temperature
Fidelity (vs. Taq) 10 X
Volume 150 μL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.