Learn More
Applied Biosystems™ TaqMan™ Microbe or ABR Assay
Product Code. 17275996 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
360 μL
Unit Size:
Each
Product Code. Quantity Unit Size
17275996 360 μL Each
1 options
Product Code. 17275996 Supplier Applied Biosystems™ Supplier No. A50133
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Applied Biosystems TaqMan Microbe Detection and TaqMan Antibiotic Resistance assays are used for quantitative real-time PCR detection of microbial species and antibiotic resistance genes.
Applied Biosystems TaqMan Microbe Detection and TaqMan Antibiotic Resistance assays consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (FAM) on the 5′ end and a minor groove binder (MGB) and non-fluorescent quencher (NFQ) on the 3′ end.
Features of TaqMan Microbe Detection and TaqMan Antibiotic Resistance assays include:
  • Easy to use—just add DNA and master mix and run the qPCR—no melt curves required
  • Specific—TaqMan assays use proprietary MGB-containing probes that can be up to 15 bases shorter than non-MGB probes, improving the specificity of the assay
  • Sensitive—TaqMan assays are ideal for measuring low-abundance microbial targets
  • Accurate—identify microbial targets with high accuracy of quantitation
  • Extensive content—over 600 predesigned microbe detection assays and more than 130 predesigned antibiotic resistance assays available
  • Gold-standard TaqMan qPCR chemistry—TaqMan assays draw on Thermo Fisher Scientific's bioinformatics assay design pipeline to help ensure high specificity and minimal cross-reactivity, even for closely related microbial species and subtypes
  • Fast delivery—assays shipped in as little as 1–2 days (see below)
Approximate ship time
  • Inventoried: 1–2 days in North America*
  • Made to order: 4–6 days in North America
All of our predesigned TaqMan gene expression assays are covered by the TaqMan Assays qPCR Guarantee.**
*Our most popular assays are stocked in North America for overnight delivery where possible. If an assay is out of stock, it may take an additional 3–5 business days for delivery.
**Subject to terms and conditions.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 360 μL