missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Yersinia pestis Detection Kit

Product Code. 10321135 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
100 reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10321135 100 reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10321135 Supplier Applied Biosystems™ Supplier No. 4382488
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ Yersinia pestis Detection Kit

Applied Biosystems™ TaqMan™ Yersinia pestis Detection Kit uses the polymerase chain reaction (PCR) to amplify a unique microorganism specific DNA target sequence and a TaqMan™ probe to detect the amplified sequence. The TaqMan™ Yersinia pestis Detection Kit provides a simple, reliable, and rapid procedure that utilizes the polymerase chain reaction (PCR) to amplify a target sequence unique to Yersinia pestis
  • Reduce sample handling with a 3-step process that uses a single, closed-tube format during the PCR step.
  • Highly sensitive TaqMan™ Real-Time PCR system enables a dramatic reduction in time required for pre-enrichment compared to conventional techniques.
  • Streamline your workflow with one standard protocol for multiple organisms.

    3 Easy Steps
    This kit has 3 easy steps: enrich samples, prepare samples, and run samples. Grow the specific pathogen from a selected sample using a standard enrichment procedure in the enrichment step. In the sample preparation step isolate the DNA for PCR, and prepare, mix and pipet the reactions. In the final step, analyze your sample by running a plate on one of our real-time PCR systems.

    A Complete Solution
    We offer a full line of TaqMan™ Pathogen Detection kits based on universal assay conditions, enabling you to run any combination of these assays together in the same sample run. In combination with our real-time PCR systems, the TaqMan™ Yersinia pestis Detection Kit offers a fully integrated, single-vendor solution to identify Yersinia pestis.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Quantity 100 reactions
    For Use With (Application) Real-Time PCR
    Reaction Speed Fast

    For Research Use Only. Not for use in diagnostic procedures.

    Name des Produkts
    Wählen Sie ein Thema

    Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.