Learn More
Applied Biosystems™ TaqMan™ Francisella tularensis Detection Kit
Alle ansehen Applied Biosystems ProdukteBeschreibung
Applied Biosystems™ TaqMan™ Francisella tularensis Detection Kit uses the polymerase chain reaction (PCR) to amplify a unique microorganism specific DNA target sequence and a TaqMan™ probe to detect the amplified sequence. The TaqMan™ Francisella tularensis Detection Kit provides a simple, reliable, and rapid procedure that utilizes the polymerase chain reaction (PCR) to amplify a target sequence unique to Francisella tularensis
- Reduce sample handling with a 3-step process that uses a single, closed-tube format during the PCR step.
- Highly sensitive TaqMan™ Real-Time PCR system enables a dramatic reduction in time required for pre-enrichment compared to conventional techniques.
- Streamline your workflow with one standard protocol for multiple organisms.
3 Easy Steps
- This kit has 3 easy steps: enrich samples, prepare samples, and run samples. Grow the specific pathogen from a selected sample using a standard enrichment procedure in the enrichment step. In the sample preparation step isolate the DNA for PCR, and prepare, mix and pipet the reactions. In the final step, analyze your sample by running a plate on one of our real-time PCR systems.
A Complete Solution
- We offer a full line of TaqMan™ Pathogen Detection kits based on universal assay conditions, enabling you to run any combination of these assays together in the same sample run. In combination with our real-time PCR systems, the TaqMan™ Francisella tularensis Detection Kit offers a fully integrated, single-vendor solution to identify Francisella tularensis
Spezifikation
Spezifikation
| Content And Storage | 300μL 10X Francisella tularensis target assay tul4, 300μL 10X francisella tularensis target assay fopA, 1,000μL negative control, two tubes 2X fast PCR master mix 1,500μL; –20/2 to 8°C |
| Description | TaqMan™ Francisella Tularensis Detection Kit, 100 rxns, Primer-Probe Detection Method, FAM™ Dye Type, F. Tularensis Target Organism Class, Tularemia Disease, Target-Specific, For Diagnostic Testing |
| Detection Method | Primer-probe |
| Label or Dye | FAM |
| Product Type | TaqMan™ Francisella Tularensis Detection Kit |
| Quantity | 100 rxn(s) |
| Target | Bacteria |
| For Use With (Application) | Pathogen Detection |
| No. of Reactions | 100 Reactions |
| Size (Final Product) | 100 rxn(s) |
For Research Use Only. Not for use in diagnostic procedures.
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.