missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Brucella spp Detection Kit

Product Code. 10249534 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
100 rxn(s)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10249534 100 rxn(s) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10249534 Supplier Applied Biosystems™ Supplier No. 4382423
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ Brucella spp. Detection Kit

Applied Biosystems™ TaqMan™ Brucella spp. Detection Kit uses the polymerase chain reaction (PCR) to amplify a unique microorganism specific DNA target sequence and a TaqMan™ probe to detect the amplified sequence.The TaqMan™ Brucella spp. Detection Kit provides a simple, reliable, and rapid procedure that utilizes the polymerase chain reaction (PCR) to amplify a target sequence unique to Brucella spp.
  • Reduce sample handling with a 3-step process that uses a single, closed-tube format during the PCR step.
  • Highly sensitive TaqMan™ Real-Time PCR system enables a dramatic reduction in time required for pre-enrichment compared to conventional techniques.
  • Streamline your workflow with one standard protocol for multiple organisms.

    3 Easy Steps
    This kit has 3 easy steps: enrich samples, prepare samples, and run samples. Grow the specific pathogen from a selected sample using a standard enrichment procedure in the enrichment step. In the sample preparation step isolate the DNA for PCR, and prepare, mix and pipet the reactions. In the final step, analyze your sample by running a plate on one of our real-time PCR systems.

    A Complete Solution
    We offer a full line of TaqMan™ Pathogen Detection kits based on universal assay conditions, enabling you to run any combination of these assays together in the same sample run. In combination with our real-time PCR systems, the TaqMan™ Brucella spp. Detection Kit offers a fully integrated, single-vendor solution to identify Brucella spp.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage 300μL 10X Brucella spp. assay mix, 1,000μL negative control, 2X fast PCR master mix 1,500μL; –20/2 to 8°C
    Description TaqMan™ Brucella spp Detection Kit, 100 rxns, Primer-Probe Detection Method, FAM™ Dye Type, Brucella spp. Target Organism Class, Brucellosis Disease, Target-Specific, For Diagnostic Testing
    Detection Method Primer-probe
    Label or Dye FAM
    Product Type TaqMan™ Brucella spp Detection Kit
    Quantity 100 rxn(s)
    For Use With (Application) Pathogen Detection
    No. of Reactions 100 Reactions
    Size (Final Product) 100 rxn(s)

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.