missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Bacillus anthracis Detection Kit

Product Code. 15840373 Shop All Applied Biosystems Products
Click to view available options
Quantity:
100 reactions
Unit Size:
100 reactions
This item is not returnable. View return policy

Product Code. 15840373

Brand: Applied Biosystems™ 4382486

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TaqMan™ Bacillus anthracis Detection Kit

Applied Biosystems™ TaqMan™ Bacillus anthracis Detection Kit uses the polymerase chain reaction (PCR) to amplify a unique microorganism specific DNA target sequence and a TaqMan™ probe to detect the amplified sequence. The TaqMan™ Bacillus anthracis Detection Kit provides a simple, reliable, and rapid procedure that utilizes the polymerase chain reaction (PCR) to amplify a target sequence unique to Bacillus anthracis
  • Reduce sample handling with a 3-step process that uses a single, closed-tube format during the PCR step.
  • Highly sensitive TaqMan™ Real-Time PCR system enables a dramatic reduction in time required for pre-enrichment compared to conventional techniques.
  • Streamline your workflow with one standard protocol for multiple organisms.

    3 Easy Steps
    This kit has 3 easy steps: enrich samples, prepare samples, and run samples. Grow the specific pathogen from a selected sample using a standard enrichment procedure in the enrichment step. In the sample preparation step isolate the DNA for PCR, and prepare, mix and pipet the reactions. In the final step, analyze your sample by running a plate on one of our real-time PCR systems.

    A Complete Solution
    We offer a full line of TaqMan™ Pathogen Detection kits based on universal assay conditions, enabling you to run any combination of these assays together in the same sample run. In combination with our real-time PCR systems, the TaqMan™ Bacillus anthracis Detection Kit offers a fully integrated, single-vendor solution to identify Bacillus anthracis
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage 300μL 10X Bacillus anthracis Target Assay pXO1 (store at -20°C), 300μL 10X Bacillus anthracis Target Assay pXO2 (store at -20°C), 1,000μL Negative Control (store at 2°C-8°C), Two Tubes 2X Fast PCR Master Mix 1,500μL (store at 2°C-8°C).
    Detection Method Primer-probe
    Format Tube
    GC-Rich PCR Performance High
    Reaction Speed Fast
    Disease Anthrax
    Label or Dye FAM
    Product Line TaqMan
    Product Type Real Time PCR Reagents
    Quantity 100 reactions
    Shipping Condition Dry Ice
    Sufficient For 100 Reactions
    Target Organism Class B. anthracis [Anthrax]
    Target Specificity Target-Specific
    For Use With (Application) Microbial Detection
    No. of Reactions 100 reactions
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.