missing translation for 'onlineSavingsMsg'
Learn More

TaqMan™ P35S Maize Quantification Kit

Product Code. 13522550 Shop All Applied Biosystems Products
Change view
Click to view available options
No. of Reactions:
48 Reactions
Unit Size:
48 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
13522550 48 Reactions 48 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13522550 Supplier ABI Applied Biosystems Supplier No. 4481972
  • ONLINE EXCLUSIVE PRICE
    £1265.00
    £1235.87 / 48 reactions
    Save £29.13
    2% Off
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ P35S Maize Quantification Kit

The Taqman™ P35S Maize Quantification Kit is a food safety testing kit used to determine the number of copies of P35S promoter present in genetically modified organisms compared to the number of copies of maize present in any food or feed sample. The benefits of the Taqman™ P35S Maize Quantification Kit over other commercially available products include:

  • Efficiency—detection of small DNA fragments through use of Taqman™ MGB probes
  • Sensitivity—quantification limit of 20 DNA copies; detection limit of 3 DNA copies (maize and P355 systems)
  • Flexibility—adapted to processed DNAs

The Taqman™ P35S Maize Quantification Kit is effective in detecting the presence of genetically modified organisms through transgenic maize quantification, based on the detection of P35S promoter. The detection success of this promoter is due to this regulatory region being present in most of the transgenic events. In addition, the Taqman™ P35S Maize Quantification Kit contains all necessary reagents for quantification and is optimized for use in all (Applied Biosystems™) real-time PCR instruments.

The development and manufacture of this kit occurs through a partnership with IMEGEN (Instituto de Medicina Genomica), which has a long history of GMO research and development.

Other related products include:

  • GMO Extraction Kit
  • TaqMan™ GMO Screening Kit
  • Taqman™ Roundup Ready Soya Quantification Kit
TRUSTED_SUSTAINABILITY

Specifications

Description TaqMan™ P35S Maize Quantification Kit, It is a Food Safety Testing Kit Used to Determine the Number of Copies of P35S Promoter Present in Genetically Modified Organisms Compared to the Number of Copies of Maize Present in any Food or Feed Sample
Detection Method Primer-probe
Kit Type TaqMan™ P35S Maize Quantification Kit
Product Type TaqMan™ P35S Maize Quantification Kit
Target P35S
No. of Reactions 48 Reactions
Sample Type Food

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.