missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ GMO Maize 35S Detection Kit, with protocol

Product Code. 12079800
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12079800 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12079800 Supplier Applied Biosystems™ Supplier No. 4327690

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Kit contains reagents needed to reliably detect the presence of genetically modified organism (GMO)-specific DNA sequences in seed, grain, processed foods, and their ingredients.

  • Contains reagents needed to reliably detect the presence of genetically modified organism (GMO)-specific DNA sequences in seed, grain, processed foods, and their ingredients
  • Universal assay detects and quantifies all approved genetically modified organisms (GMOs) in Europe, and the vast majority of GMOs approved in other countries
  • Fluorogenic 5' nuclease assay using TaqMan™ probes provides accurate and reproducible quantitative results
  • Comprehensive controls provide high confidence in results
  • Innovative GMO quantitation software allows users to quickly and easily determine the percentage of GMO content
  • Offers the complete solution including sample preparation, instruments, reagents, analysis software, technical support, instrument service, and GMO expertise
TRUSTED_SUSTAINABILITY

Specifications

Detection Method Primer-Probe
For Use With (Equipment) 7300 System, 7500 System, AB 7900 HT 384-Well System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
Content And Storage Each kit contains TaqMan GMO Mix, AmpliTaq Gold™ DNA Polymerase, positive and negative controls. Each kit contains enough regents for 100 assays.

Store all components at -25 to -15°C.
Shipping Condition Dry Ice
Sample Type Food, Seed
Target Organism Class Maize, Genetically Modified
Quantity 1 Kit
Product Line TaqMan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.