missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Fast Reagent Starter Kit

Product Code. 15871572 Shop All Applied Biosystems Products
Click to view available options
Quantity:
125 reactions
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15871572

Brand: Applied Biosystems™ 4352407

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed to detect and quantify Eukaryotic 18S rRNA sequence in cDNA samples using the TaqMan 5' nuclease assay, in fast thermal cycling mode.

The Eukaryotic 18S rRNA TaqMan™ Assay consists of a 20X mix of two unlabeled PCR primers and a FAM™ dye labeled 18S TaqMan™ MGB probe. Used With: 7900HT Fast Real-Time PCR System with Fast 96-well Block Module, 7900HT Fast Real-Time Service Upgrade, 7900HT System Fast 96-well Block Upgrade Kit.

Order Info

Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage This kit contains TaqMan™ Fast Real-Time PCR Universal Master Mix (2X), no AmpErase™ UNG (1 tube) , Eukaryotic 18S rRNA TaqMan™ Assay (20X mix of two unlabeled PCR primers and a FAM™ dye-labeled 18S TaqMan™ MGB probe) (Assay ID Hs99999901_s1; Part Number 4331182), and human Raji cDNA.

Store the PCR mix at 2° to 8°C
Store the rest of the kit contents at -15° to -25°C.
Detection Method Primer-probe
PCR Method qPCR
Polymerase AmpliTaq DNA Polymerase
Reaction Speed Fast
For Use With (Equipment) 7500 Fast System, 7900HT Fast System, AB 7900HT Fast Mode
Passive Reference Dye ROX (Pre-mixed)
Product Line TaqMan™
Product Type PCR Reagents
Quantity 125 reactions
Shipping Condition Dry Ice
Sufficient For 125 Reactions
Target 18S rRNA
Target Specificity Target-Specific
For Use With (Application) Gene Expression
Hot Start No
No. of Reactions 125 Reactions
Sample Type DNA (Genomic), cDNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.