missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Copy Number Reference Assay, mouse, Tfrc
GREENER_CHOICE

Product Code. p-7076280 Shop All Applied Biosystems Products
missing translation for 'orderingAttributeHoverText'
No. of Reactions:
3000 Reactions
750 Reactions
missing translation for 'unitSize'
750 reactions
750µL
This item is not returnable. View return policy

Product Code. 15305089

missing translation for 'mfr': Applied Biosystems™ 4458367

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse TaqMan™ Copy Number Reference Assays are run with predesigned, Custom Plus, and Custom TaqMan™ Copy Number Assays in a duplex, real-time PCR reaction to detect and measure copy number variations in the mouse genome.

Mouse TaqMan® Copy Number Reference Assays are run with predesigned, Custom Plus, and Custom TaqMan® Copy Number Assays in a duplex, real-time PCR reaction to detect and measure copy number variations in the mouse genome. Mouse TaqMan® Copy Number Reference Assays are also compatible with the common vector marker and reporter gene predesigned TaqMan® Copy Number Assays. Two options for genes that can be used as endogenous reference genes in mice are offered: Tfrc and Tert. TaqMan® Copy Number Reference Assays are designed to unique genomic sequences in the mouse reference genome assembly (NCBI Build 37/mm9) and are required for relative quantitation of copy number targets.

TaqMan® Copy Number Assays quantitate the gene of interest and normalize to an endogenous reference gene known to be present in two copies in a diploid genome. Please note that TaqMan® Copy Number Reference Assays are species-specific.

Tfrc: The Standard Reference Gene Option
TaqMan® Copy Number Reference Assays have a VIC® dye-labeled TAMRA™ probe and reference sequence-specific forward and reverse primers. The assays are not primer limited.

TaqMan® Copy Number Reference Assay, Mouse, Tfrc is recommended as the standard reference assay for copy number analysis in mice. This assay detects the transferrin receptor gene (Tfrc) on chromosome 16, cytoband 16qB3. The assay location is chr.16:32626732 on NCBI build 37. It has a 91 bp amplicon that maps within exon 17 of the Tfrc gene.

TaqMan® Copy Number Reference Assay, Mouse, Tert is an alternative reference assay; it is recommended in the event that the Tfrc assay functions poorly with a sample because of chromosomal aberrations or other issues. The assay targets the telomerase reverse transcriptase (Tert) gene on chromosome 13, cytoband 13qC1. The assay location is chr.13:73778992 on NCBI build 37. It has a 96 bp amplicon that maps within intron 8 of the Tert gene.

Simplest Workflow Available
TaqMan® Copy Number Assays have the simplest workflow of all currently available CNV analysis methods. The test assay (FAM™ dye labeled), the reference assay (VIC® dye labeled), your sample DNA, and TaqMan® Master Mix are combined and then run on an Applied Biosystems® Real-Time PCR System using the standard TaqMan® Copy Number Assay protocol. On average, set-up to primary analysis takes only 3-4 hours (including an approximately 2 hour PCR run).

Powerful Data Analysis Software
CopyCaller® Software was developed specifically for TaqMan® Copy Number Assay data analysis. This free, easy-to-use software utilizes a graphical interface that quickly calculates the possible copy numbers for a set of samples in a run. It also gives a confidence value for each copy number call, and has outlier functionality.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.

TRUSTED_SUSTAINABILITY

Specifications

3'Primer Modification None
5'Primer Modification None
Concentration 20X
Content And Storage Control Primer-Probe Set(s)
Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance Low
PCR Method qPCR
Reaction Speed Standard
Gene Symbol TFRC
Green Features Sustainable packaging
Internal Probe Modification VIC (5'), TAMRA Quencher (3')
Label or Dye VIC
Product Line TaqMan
Product Type Copy Number Reference Assay
Quantity 3,000 reactions
Shipping Condition Dry Ice
Species Mouse
Target Tfrc (Transferrin Receptor)
No. of Reactions 3000 Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.