missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Array, Plate 48
Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 Plate
Unit Size:
Each
Description
Custom TaqMan™ Plates are cost-effective and convenient 96-well formats for TaqMan™ Gene Expression Assays. Choose from over 50,000 inventoried human, mouse, rat, canine, and rhesus macaque assays, and from five plate formats - single assays, or assays in duplicate or triplicate.
Specifications
Specifications
| Array | 6 x 8; 48-well Plate; Format 48 |
| Content And Storage | Store at room temperature. |
| Description | TaqMan™ Array Plates, 20 μL Reaction, 48 Format (Standard), Used for PCR and real-time PCR, real time PCR (qPCR), gene expression analysis and genotyping, real time PCR-based gene expression profiling |
| Format | Standard |
| Quantity | 1 Plate |
| For Use With (Application) | Gene Expression, Genotyping |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction