missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array Human Phosphodiesterase Panel

Product Code. 11630109 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 cards
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11630109 4 cards Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11630109 Supplier Applied Biosystems™ Supplier No. 4378705
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ Array Human Phosphodiesterase Panel

The Applied Biosystems™ TaqMan™ Human Phosphodiesterase Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of inflammation genes important in drug discovery. This product is designed with 48 TaqMan™ Gene Expression Assays arrayed in 384 wells and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System and sold in packs of 4. The array contains 43 human assays and five human controls. Orthologous genes to the human panel are in the Rat Phosphodiesterase Panel with two exceptions: Pde6g and Gdpd4 are missing and two added controls (Arbp and Ywhaz) are in the rat panel.

The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
TRUSTED_SUSTAINABILITY

Specifications

Type Array Human Phosphodiesterase Panel
Format 384-well Microfluidic Card
Species Human
Content And Storage Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored at 4°C.
Form Dried Down
Includes TaqMan Assay for One 2 uL Reaction
Internal Probe Modification MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or Dye FAM
Product Line TaqMan
Quantity 4 cards
Reaction Speed Standard
Research Category Cell Cycle & Proliferation, Neurobiology, Toxicology & Drug Metabolism, Immunology, Oncology
Target Gene Includes five distinct phosphodiesterase families
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.