Learn More
Applied Biosystems™ TaqMan™ Array, Human Osteogenesis, Fast 96-well
TaqMan™ Array, Human Osteogenesis, Fast 96-well
Brand: Applied Biosystems™ 4418741
184.30 GBP valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00450kg
Description
These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The panel of assays in the TaqMan™ Array 96-well Human Osteogenesis Plate targets genes associated with osteoblast determination and differentiation, including the RUNX2 transcription factor and growth factors with induce osteoprogenetor to differentiation and which guide osteoblast function (bone morphogenetic proteins, etc). The panel also includes genes involved in the physiological processes of bone and tooth formation, mineralization and maintenance, including collagens, enamelin and matrix metalloproteinasesSpecifications
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or at 4°C. | |
96-well plate | |
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') | |
TaqMan™ | |
Human | |
1 x 10 μL Reactions |
Dried Down | |
Fast | |
FAM | |
Cell Cycle & Proliferation, Signal Transduction, Biomarker-Related Pathway(s), Neurobiology, Development | |
Gene Expression |
For Research Use Only. Not for use in diagnostic procedures.