Learn More
Applied Biosystems™ TaqMan™ Array Human Neurotransmitters
Shop All Applied Biosystems ProductsDescription
The panel of assays in this plate targets genes associated with several common groups of neurotransmitters and neurotransmitter receptors: Acetylcholines (CHRM and CHRN); Dopamine receptors; GABA receptors; glycine receptors; glutamate receptors and 5-HT (serotonin) receptors. Genes involved in neurotransmitter synthesis, transport, and degradation are represented in the panel.
Gene Signature Plates are 96-well plates that are pre-configured with the most appropriate TaqMan Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment.
TaqMan Array Plates contain the same TaqMan Gene Expression Assays that have been available for years in a convenient, dried, 96-well format. Tests have found no difference in performance between TaqMan Assays supplied in the liquid format and assays dried into a TaqMan Array Plate. Ideal for projects such as validation of microarray and RNAi data, and pathway studies, the plates include enough TaqMan Assay in each well for one 20μL reaction.
Each pack of TaqMan Array Plates is shipped with a CD containing the plate map along with details of each TaqMan Assay. This information is imported into the real-time PCR instrument to enable proper labeling of experimental data and easy access to Assay ID numbers for simple reordering.
- Affordable; minimum order of three plates; ideal for small, medium or large-sized projects
- Fast delivery; on your bench in days
- Convenient; dried PCR primer and TaqMan™ probe sets supplied in a 96- well plate; just add master mix and your sample
- TaqMan Array Plates have been optimized for use with TaqMan Gene Expression Master Mix, but are also compatible with TaqMan Universal PCR Master Mix
- For studies using cultured cells, TaqMan Array Plates are also compatible (and recommended) with the TaqMan Gene Expression Cells-to-Ct™ Kit
- TaqMan Array Plates must be run on a real-time PCR instrument with 96-well plate capability
- The plates have been validated for use on Applied Biosystems™ 7000, 7300, 7500 and 7900HT Fast Real-Time PCR Systems using standard 96-well blocks, standard PCR cycling conditions, and 20μL reaction
Specifications
Specifications
| Content And Storage | Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 20 μL reaction. The plates can be stored at room temperature or at 4°C. |
| Form | Dried Down |
| Format | 96-well plate |
| Reaction Speed | Standard |
| Internal Probe Modification | MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') |
| Label or Dye | FAM |
| Product Line | TaqMan |
| Purification Method | Standard |
| Quantity | 1 standard plate |
| Research Category | Biomarker-Related Pathway(s), Neurobiology |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.