Learn More
Applied Biosystems™ TaqMan™ Array Human Inflammation Panel
The Applied Biosystems™ TaqMan™ Human Inflammation Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of inflammation genes important in drug discovery.
Brand: Applied Biosystems™ 4378707
Code : NEW
Additional Details : Weight : 0.00450kg
Description
- An efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of inflammation genes important in drug discovery
- Designed with 96 TaqMan™ Gene Expression Assays arrayed in 384 wells and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System and is sold in packs of 4
- TaqMan™ Gene Signature Arrays for human inflammation contain over 90 genes that are, or have been, studied as targets for a range of inflammatory diseases
- The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics
- Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators
Specifications
Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored at 4°C. | |
384-well microfluidic card | |
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') | |
TaqMan™ | |
Human | |
Gene Expression |
Dried Down | |
Standard | |
FAM | |
Signal Transduction, Oncology, Biomarker-Related Pathway(s), Extracellular Matrix & Cell Adhesion, Inflammation | |
Over 90 genes that are targets for a range of inflammatory diseases | |
1 x 2 μL Reactions |
For Research Use Only. Not for use in diagnostic procedures.