missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array Human Apoptosis Panel

Product Code. 11690099 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 Cards
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11690099 4 Cards Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11690099 Supplier Applied Biosystems™ Supplier No. 4378701
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ Array Human Apoptosis Panel

The Applied Biosystems™ TaqMan™ Human Apoptosis Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of targets known to have implications in programmed cell death. The study of apoptosis is important in cancer, HIV, and other disease research. This product is designed with 96 TaqMan™ Gene Expression Assays (93 targets & 3 controls) loaded in singletons into the array’s 384 wells and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System. This product is packaged in 1 box containing 4 cards.

The TaqMan™ Human Apoptosis Array contains assays for 93 human genes in addition to 3 endogenous controls (18S, ACTB, GAPDH). The 93 genes are categorized in multiple target classes or pathways. The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
TRUSTED_SUSTAINABILITY

Spezifikation

Type TaqMan™ Array Human Apoptosis Panel
Format 384-well Microfluidic Card
Content And Storage 4°C
For Use With (Application) Cell viability, proliferation and function, oncology and genetic disease research, pharma and biopharma, real-time PCR (qPCR), real-time PCR-based gene expression profiling, target and lead identification and validation
Includes TaqMan™ array well contains a single TaqMan™ assay for one 2μL reaction
Quantity 4 Cards

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.