missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array, Format 96b

Product Code. 15571034 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15571034 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15571034 Supplier Applied Biosystems™ Supplier No. 4342261
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TaqMan™ Array, Format 96b

The Custom TaqMan™ Array Card is a 384-well microfluidic card designed to perform 384 simultaneous real-time PCR reactions without using liquid-handling robots or multichannel pipettors. Select target assays from thousands of inventoried TaqMan™ Gene Expression Assays.
TRUSTED_SUSTAINABILITY

Specifications

Type Custom Gene Expression TaqMan™ Array Cards
Format 96B
Content And Storage Room Temperature
No. of Samples 2
Quantity 1 Ea.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.