missing translation for 'onlineSavingsMsg'
Learn More

TaqMan™ Advanced miRNA Human A Card

Product Code. 17193089 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
384 reactions
Unit Size:
384 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17193089 384 reactions 384 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17193089 Supplier ABI Applied Biosystems Supplier No. A34714

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Applied Biosystems TaqMan Advanced miRNA Human A Card provides the convenience of pre-spotted TaqMan Advanced miRNA assays in a 384-well microfluidic card format.

This product contains one micro-fluidic card, enabling accurate quantitation of 384 unique microRNAs in as little as five hours. Four TaqMan Advanced miRNA Assay endogenous controls are included in the card to aid in data normalization. Details about the included assays are available in the Target List file. The assays can detect and quantify the mature form of the miRNA from 1–10 ng of total RNA from tissue.

Note: The TaqMan Advanced miRNA cDNA Synthesis Kit is required for preparation of the cDNA template that is used with the TaqMan Advanced miRNA assays.

The kit enables the analysis of:

  • Multiple miRNAs from a single amplified sample
  • Samples that are limited in quantity, including serum, plasma, or other biological fluids

Key features include:

  • Universal reverse transcription (RT)—one RT step for all TaqMan Advanced miRNA assays
  • Sensitive—detect as few as 60 copies of input miRNA
  • Specific—detect only mature miRNA and distinguish between highly homologous miRNAs
  • Small sample input—detect and quantify mature miRNA from as little as 1–10 ng of total RNA or 2 μL of plasma or serum
  • Versatile—compatible with tissue and biofluids, including serum and plasma

TaqMan Advanced miRNA cards provide all the advantages of TaqMan Advanced miRNA assays in a convenient, pre-configured microfluidic card, minimizing the experimental variability and effort required to run 384 TaqMan Advanced miRNA assays in parallel. With the TaqMan Advanced miRNA cDNA Synthesis Kit, TaqMan Advanced miRNA cards enable a comprehensive expression profile consistent with miRBase v21, providing the ideal miRNA profiling solution.

TaqMan Advanced miRNA cards are compatible with the following Applied Biosystems qPCR instruments: Viia7, QuantStudio 7 Flex, and QuantStudio 12k Flex.

TaqMan Advanced miRNA cards require the use of the TaqMan Advanced cDNA Synthesis Kit, which uses a universal RT chemistry to prepare the cDNA template for use with TaqMan Advanced miRNA assays for detection and quantification of mature human miRNAs in biological samples.

These plates are not compatible with the first-generation TaqMan MicroRNA cDNA Synthesis Kit, which uses an miRNA-specific reverse transcription chemistry.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage One 384-well array

The TaqMan miRNA card is shipped at ambient temperature. Upon arrival, store at 2–8°C and keep protected from light.
Detection Method Primer-probe
Format 384-well microfluidic card
GC-Rich PCR Performance High
PCR Method qPCR
Reaction Speed Fast or Standard
Label or Dye 2 Color
Product Line TaqMan
Product Type Advanced miRNA Human A Card
Quantity 384 reactions
Research Category miRNA
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Species Human
For Use With (Application) miRNA analysis
Volume 2 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.