missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Taq-&LOAD™ PCR Mastermix

Product Code. 11447010 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 x 100U
100U
Unit Size:
100 units
1000 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447010 100U 100 units
11457010 10 x 100U 1000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11447010 Supplier MP Biomedicals™ Supplier No. 11EPTAL100
  • ONLINE EXCLUSIVE PRICE
    £130.00
    £121.80 / Pack of 100
    Save £8.20
    6% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This mastermix allows direct loading after the PCR on the agarose gel without the need of loading dye.

Taq-&LOAD PCR Mastermix is a 5X concentrated solution containing MP Biomedicals recombinant Taq DNA Polymerase, dNTPs, MgCl2, incubation and loading buffer (includes a compound increasing the density of the sample and a red/purple dye). The included red/purple dye migrates at 400 bp on a 1.2% agarose gel.

Specifications

Concentration 5X
Content And Storage Recombinant Taq DNA Polymerase, dNTPs, MgCl2, incubation and loading buffer. Store at -20°C
Format Tube
Gel Compatibility Agarose
Label or Dye Red-purple
Quantity 100U
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.