missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Taq DNA Polymerase

Product Code. 16062545
Click to view available options
Quantity:
1000 units
2500 units
500 units
Unit Size:
1000 units
2500 units
500 units
This item is not returnable. View return policy

Product Code. 16062545

Brand: Abnova™ D0010.1000Units

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Taq DNA polymerase catalyzes 5'-3' synthesis of DNA.

  • The enzyme has been proved not having the 3′-5′ exonuclease activity.
  • The enzyme has proven to have high amplification yield, be stable at high temparetaure.

Specifications

Concentration 5 U/μL
Content And Storage Store at -20°C Guaranteed stable for 6 months when properly stored
Quantity 1000 units
For Use With (Application) Standard PCR
Storage Buffer 20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.