missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Taq DNA Polymerase

Taq DNA polymerase catalyzes 5′-3′ synthesis of DNA.

Brand:  Abnova™ D0010.500Units

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16082545

  • £90.00 / 500 units
Estimated Shipment: 26-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • The enzyme has been proved not having the 3′-5′ exonuclease activity.
  • The enzyme has proven to have high amplification yield, be stable at high temparetaure.
Specifications

Specifications

Store at -20°C Guaranteed stable for 6 months when properly stored
Standard PCR
20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol
500 units
5 units/µL
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers