missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Taq CORE Kit 25

Product Code. 11427040 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 x 250U
12 x 600U
5 x 1000U
Unit Size:
2500 units
5000 units
7200 units
This item is not returnable. View return policy

Product Code. 11427040

Brand: MP Biomedicals™ 11EPTQK600

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Taq DNA Polymerase 5U/μL, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 25mM dNTP Mix

Supplied with standard reaction buffers

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Taq DNA Polymerase 5U/μL, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 25mM dNTP Mix. Store at -20°C
Quantity 12 x 600U
For Use With (Application) Standard PCR
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.