missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Taq CORE Kit 25

Supplied with standard reaction buffers

Brand:  MP Biomedicals™ 11EPTQK25L

Code : 80

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11407040

  • £2553.54 / 5000 units
Estimated Shipment: 16-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: Taq DNA Polymerase 5U/μL, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 25mM dNTP Mix

Description

Description

Specifications

Specifications

Taq DNA Polymerase 5U/μL, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 25mM dNTP Mix. Store at -20°C
5 x 1000U
Standard PCR
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers