missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAPT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA524318
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with chicken based on sequence homology.
This gene encodes a highly conserved, putative transmembrane protein. A mutation in the mouse ortholog of this gene results in homeotic, posterior-to-anterior transformations of the axial skeleton which are similar to the phenotype of mouse homeobox C8 gene mutants. This gene is proposed to function downstream of homeobox C8 to transduce extracellular patterning information during axial skeleton development. An alternatively spliced transcript variant encoding a substantially different isoform has been described, but its biological validity has not been determined.Specifications
TAPT1 | |
Polyclonal | |
Unconjugated | |
TAPT1 | |
4932414K18Rik; CMVFR; Cytomegalovirus partial fusion receptor; OCLSBG; TAPT1; transmembrane anterior posterior transformation 1; transmembrane anterior posterior transformation protein 1; Transmembrane anterior posterior transformation protein 1 homolog | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
202018, 231225 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q4VBD2, Q6NXT6 | |
TAPT1 | |
KLH conjugated synthetic peptide between 530-558 amino acids from the C-terminal region of human TAPT1 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |