missing translation for 'onlineSavingsMsg'
Learn More

TAPS, 99%

Product Code. 11473348
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473348 25 g 25g
11483348 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473348 Supplier Thermo Scientific Alfa Aesar Supplier No. A17754.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29915-38-6 | C7H17NO6S | 243.27 g/mol

A zwitterionic Good's Buffer

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29915-38-6
Molecular Formula C7H17NO6S
Molecular Weight (g/mol) 243.27
MDL Number MFCD00007538
InChI Key YNLCVAQJIKOXER-UHFFFAOYSA-N
Synonym taps, 3-1,3-dihydroxy-2-hydroxymethyl propan-2-yl amino propane-1-sulfonic acid, unii-y5dc3in066, n-tris hydroxymethyl methyl-3-aminopropanesulfonic acid, 2-hydroxy-1,1-bis hydroxymethyl ethyl amino-1-propanesulfonic acid, 3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino-1-propanesulfonic acid, taps buffer, 1-propanesulfonic acid, 3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino
PubChem CID 121591
IUPAC Name 3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonic acid
SMILES OCC(CO)(CO)NCCCS(O)(=O)=O

Specifications

Melting Point 239°C to 241°C (decomposition)
Quantity 25 g
Formula Weight 243.28
Percent Purity 99%
Chemical Name or Material TAPS

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.