missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TAPS, 99+%, for biochemistry

Product Code. 10317901 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10317901 100 g Glass bottle 100g
10716623 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10317901 Supplier Thermo Scientific Chemicals Supplier No. 327801000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 29915-38-6 | C7H17NO6S | 243.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29915-38-6
Molecular Formula C7H17NO6S
Molecular Weight (g/mol) 243.27
MDL Number MFCD00007538
InChI Key YNLCVAQJIKOXER-UHFFFAOYSA-N
Synonym 3-[Tris(hydroxymethyl)methylamino]-1-propanesulfonic acid
PubChem CID 121591
IUPAC Name 3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonic acid
SMILES OCC(CO)(CO)NCCCS(O)(=O)=O

Specifications

Absorbance 0.04 max. at 260nm
Absorbance Conditions (0.1M aq. soln.)
Melting Point 240.0°C
Color White
pH 5 to 6.5 (1% aq. soln. at 25°C)
Physical Form Powder
Quantity 100 g
Flash Point >110°C
Assay Percent Range 99+%
Infrared Spectrum Authentic
Linear Formula (HOCH2)3CNH(CH2)3SO3H
Packaging Glass bottle
Merck Index 15, 9191
Solubility Information Solubility in water: ca 121.6g/L (20°C).
Formula Weight 243.27
Percent Purity ≥99%
Grade Biochemical
Chemical Name or Material TAPS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.