missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ TAPS
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
- Application Notes: TAPS is utilized in capillary electrophoresis, e.g., DNA and DNA-dye complexes.
- Usage Statement: Unless specified otherwise, products are for research or further manufacturing use only, not for direct human use.
Specifications
Specifications
| Chemical Name or Material | N-[Tris(hydroxymethyl)methyl]-3-aminopropanesulfonic acid |
| CAS | 29915-38-6 |
| Color | White |
| pH | 5.5 to 6.5 (1% aq. soln.), useful pH range 7.7 to 9.1 |
| Physical Form | Powder |
| Quantity | 25 g |
| Molecular Formula | C7H17NO6S |
| Synonym | N-[Tris(hydroxymethyl)methyl]-3-aminopropanesulfonic acid; [(2-Hydroxy-1,1-bis(hydroxymethyl)ethyl)amino]-1-propanesulfonic acid; 1-bis(hydroxymethyl)ethyl)amino]-1-propanesulfonic acid; 3-[Tris(hydroxymethyl) methylamino]-1-propane sulfonic acid |
| Solubility Information | Water Soluble |
| Molecular Weight (g/mol) | 243.274 g/mol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction