missing translation for 'onlineSavingsMsg'
Learn More

Taprenepag, MedChemExpress

Product Code. 16488837
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488837 5 mg 5mg
16466248 10 mg 10mg
16442377 50 mg 50mg
16416596 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16488837 Supplier Medchem Express Supplier No. HY14899
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Taprenepag (CP-544326) is a potent and selective prostaglandin EP(2) agonist with IC50s of 10 and 15 nM for human and rat EP2, respectively. Taprenepag shows selectivity for EP2 over other EP receptors (IC50s>3200 nM for EP1, EP3, and EP4) and a panel of 37 G protein-coupled receptors.

Taprenepag (CP-544326) is a potent and selective prostaglandin EP(2) agonist with IC50s of 10 and 15 nM for human and rat EP2, respectively. Taprenepag shows selectivity for EP2 over other EP receptors (IC50s>3200 nM for EP1, EP3, and EP4) and a panel of 37 G protein-coupled receptors.

Specifications

Chemical Name or Material Taprenepag
CAS 752187-80-7
Color White
Purity Grade Notes Research
Molecular Formula C24H22N4O5S
Quantity 5 mg
Synonym CP-544326
Solubility Information DMSO : ≥ 100 mg/mL (208.98 mM)
SMILES O=S(C1=CN=CC=C1)(N(CC2=CC(OCC(O)=O)=CC=C2)CC3=CC=C(N4C=CC=N4)C=C3)=O
Molecular Weight (g/mol) 478.52
Formula Weight 478.52
Percent Purity 99.54%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.