missing translation for 'onlineSavingsMsg'
Learn More

Tapinarof, MedChemExpress

Product Code. 16494258
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16494258

Brand: Medchem Express HY109044

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tapinarof (WBI-1001) is a natural aryl hydrocarbon receptor (AhR) agonist with an EC50 of 13 nM. Tapinarof resolves skin inflammation in mice.

Tapinarof (WBI-1001) is a natural aryl hydrocarbon receptor (AhR) agonist with an EC50 of 13 nM. Tapinarof resolves skin inflammation in mice.

Specifications

Chemical Name or Material Tapinarof
CAS 79338-84-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H18O2
Quantity 10 mg
Synonym WBI-1001 Benvitimod GSK2894512
Solubility Information DMSO : 100 mg/mL (393.21 mM; Need ultrasonic)
SMILES OC1=C(C(C)C)C(O)=CC(/C=C/C2=CC=CC=C2)=C1
Molecular Weight (g/mol) 254.32
Formula Weight 254.32
Percent Purity 99.17%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.