missing translation for 'onlineSavingsMsg'
Learn More

Tapinarof, MedChemExpress

Product Code. 16410597
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16410597 50 mg 50mg
16494258 10 mg 10mg
16423016 25 mg 25mg
16406987 5 mg 5mg
16405196 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16410597 Supplier Medchem Express Supplier No. HY109044
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tapinarof (WBI-1001) is a natural aryl hydrocarbon receptor (AhR) agonist with an EC50 of 13 nM. Tapinarof resolves skin inflammation in mice.

Tapinarof (WBI-1001) is a natural aryl hydrocarbon receptor (AhR) agonist with an EC50 of 13 nM. Tapinarof resolves skin inflammation in mice.

Specifications

Chemical Name or Material Tapinarof
CAS 79338-84-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H18O2
Quantity 50 mg
Synonym WBI-1001 Benvitimod GSK2894512
Solubility Information DMSO : 100 mg/mL (393.21 mM; Need ultrasonic)
SMILES OC1=C(C(C)C)C(O)=CC(/C=C/C2=CC=CC=C2)=C1
Molecular Weight (g/mol) 254.32
Formula Weight 254.32
Percent Purity 99.17%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.