Learn More
Fisherbrand™ Tapered PTFE Stir Bars
Brand:  Fisherbrand™ 001.1950
Product Code. 10323692
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Stable, general-purpose magnetic stir bar
  • Tapered ends provide similar action as oval or pivot ring stirrers
  • Autoclavable