Learn More
Invitrogen™ TAPBPL Monoclonal Antibody (OTI1C9)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA526150
Product Code. 15975744
  • £487.00 / 100µL
Description
TAPBPL Monoclonal Antibody for Western Blot, IHC (P)
Tapasin, or TAPBP, is a member of the variable-constant Ig superfamily that links major histocompatibility complex (MHC) class I molecules to the transporter associated with antigen processing in the endoplasmic reticulum (ER). The TAPBP gene is located near the MHC complex on chromosome 6p21.3. TAPBPL is a member of the Ig superfamily that is localized on chromosome 12p13.3, a region somewhat paralogous to the MHC.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TAPBPL
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BX59
TAPBPL
Full length human recombinant protein of TAPBPL produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI1C9
Unconjugated
TAPBPL
BC017613; TAP binding protein like; TAP binding protein related; TAP binding protein-like; tapasin-like; TAPASIN-R; tapasin-related protein; tapasin-related protein-like; TAP-binding protein-like; TAP-binding protein-related protein; Tapbpl; Tapbplr; TAPBPL-R; TAPBPR; TAPBP-R
Mouse
Affinity Chromatography
RUO
55080
-20°C, Avoid Freeze/Thaw Cycles