missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tapasin Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542731
This item is not returnable.
View return policy
Description
Peptide sequence: GEAPPELLCL VSHFYPSGGL EVEWELRGGP GGRSQKAEGQ RWLSALRHHS Sequence homology: Cow: 86%; Human: 100%; Pig: 86%; Sheep: 86%.
This gene encodes a transmembrane glycoprotein which mediates interaction between newly assembled major histocompatibility complex (MHC) class I molecules and the transporter associated with antigen processing (TAP), which is required for the transport of antigenic peptides across the endoplasmic reticulum membrane. This interaction is essential for optimal peptide loading on the MHC class I molecule. Up to four complexes of MHC class I and this protein may be bound to a single TAP molecule. This protein contains a C-terminal double-lysine motif (KKKAE) known to maintain membrane proteins in the endoplasmic reticulum. This gene lies within the major histocompatibility complex on chromosome 6. Alternative splicing results in three transcript variants encoding different isoforms.
Specifications
| Tapasin | |
| Polyclonal | |
| Unconjugated | |
| TAPBP | |
| D17Wsu91e; DADB-159G18.7; NGS17; NGS-17; similar to Tapasin precursor (TPSN) (TPN) (TAP-binding protein) (TAP-associated protein) (NGS-17); TAP binding protein; TAP binding protein (tapasin); TAPA; tapasin; TAP-associated protein; TAP-binding protein; Tapbp; TPN; TPSN | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6892 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O15533 | |
| TAPBP | |
| Synthetic peptide directed towards the C-terminal of human TAPBP (aa 307-356). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction